Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Mouse anti-Human, Rat, Alexa Fluor 700, Clone: 7H12, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147483AF700
Description
Synaptophysin Monoclonal specifically detects Synaptophysin in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Synaptophysin | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SYP | |
Purified recombinant fragment of human Synaptophysin expressed in E. coli. [UniProt# P08247] | |
0.1 mL | |
Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers | |
6855 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
7H12 | |
Western Blot, ELISA, Immunohistochemistry-Paraffin | |
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only