Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Rabbit anti-Chicken, Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc SYN112AP

Description
Synaptophysin Polyclonal antibody specifically detects Synaptophysin in Chicken, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western BlotSpecifications
Synaptophysin | |
Polyclonal | |
Unconjugated | |
Syp | |
A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1 | |
Synthetic peptide taken within amino acid region 250-350 on human Synaptophysin protein. | |
200 μL | |
Primary | |
Chicken, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P07825, P08247, Q62277 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20977, 24804, 465628, 6855 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction