Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Rabbit anti-Human, Clone: SYP/4503R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30732520UG
Description
Synaptophysin Monoclonal specifically detects Synaptophysin in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Synaptophysin | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
Recombinant fragment (around aa 224-313) of human Synaptophysin protein (exact sequence is proprietary) | |
20 μg | |
Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers | |
6855 | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin) | |
SYP/4503R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title