Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptophysin Rabbit anti-Human, Clone: SYP/4503R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Synaptophysin
Clone SYP/4503R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122124
View Documents
Novus Biologicals
NBP30732520UG
20 μg
Each for $159.82
Add to cart
 
NB122123
View Documents
Novus Biologicals
NBP30732510UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Synaptophysin Monoclonal antibody specifically detects Synaptophysin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Synaptophysin
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin
Recombinant fragment (around aa 224-313) of human Synaptophysin protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SYP/4503R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Angiogenesis, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Stem Cell Markers
10 mM PBS with 0.05% BSA
6855
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit