Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptophysin Sheep anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
$177.31 - $436.00
Specifications
Antigen | Synaptophysin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10098781
|
Novus Biologicals
NB10098781 |
0.1 mL |
Each for $436.00
|
|
NB017949
|
Novus Biologicals
NB100987810.025ML |
0.025 mL |
Each for $177.31
|
|
Description
Synaptophysin Polyclonal specifically detects Synaptophysin in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Synaptophysin | |
Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000 | |
Polyclonal | |
Antisera | |
RUO | |
Mouse, Rat | |
Q62277 | |
6855 | |
A synthetic peptide from internal region of mouse Synaptophysin (SYP) conjugated to an immunogenic carrier protein was used as the antigen. | |
Primary | |
Reconstitute with 0.1 ml sterilized water to desired concentration. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Sheep | |
Membrane Vesicle Markers, Stem Cell Markers | |
Whole antisera with No Preservative | |
Major synaptic vesicle protein p38, MRX, MRXSYP, synaptophysin | |
SYP | |
IgG | |
Unpurified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title