Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptopodin Rabbit anti-Human, Mouse, Rat, Alexa Fluor™ 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP239100AF488

Catalog No. NP23910A488


Add to cart

Description

Description

Synaptopodin Polyclonal antibody specifically detects Synaptopodin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

Synaptopodin
Alexa Fluor 488
50 mM sodium borate with 0.05% sodium azide
KIAA1029, synaptopodin
Rabbit
IgG
0.1 mL
Primary
11346.0
Human, Mouse, Rat
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
SYNPO
Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SYNPO. The immunogen is located within the last 50 amino acids of SYNPO. Amino Acid Squence: SPSRAWSPRAKQAPRPS
Peptide affinity purified
Store at 4°C in the dark.
Polyclonal
At least three isoforms of SYNPO are known to exist; this antibody will detect all three isoforms. SYNPO antibody is predicted to not cross-react with other SYNPO family member proteins.
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit