Learn More
Synaptopodin Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP146529
 View more versions of this product
Catalog No. NBP146529
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Synaptopodin Polyclonal antibody specifically detects Synaptopodin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Synaptopodin
LYOPH
Western Blot 1: 300-1: 2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin
Q8N3V7, Q8CC35, Q9Z327
KIAA1029, synaptopodin
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specific for SYNPO.
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Whole antisera with No Preservative
Antisera
SYNPO
A synthetic peptide from the c-terminal region of human Synaptopodin (SYNPO) conjugated to an immunogenic carrier protein was used as the antigen.
Unpurified
RUO
Primary
11346
Reconstitute with 0.1 ml sterilized water to desired concentration.