Learn More
BioworldSupplier Diversity Partner SYNAPTOSOMAL-ASSOCIATED PRO DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220607051
SYNAPTOSOMAL-ASSOCIATED PROTEIN 25KDA, C.ELEGANS (RECOMBINANT): Recombinant Synaptosomal-associated protein 25kDa, C.elegans (SNAP-25) Source: E.Coli Background: SNAP-25 is a presynaptic plasma membrane protein essential for synaptic...
Catalog No. 50-253-806