Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptotagmin 1 Rabbit anti-Human, Mouse, Rat, FITC, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc SYT1FITC

Description
Synaptotagmin 1 Polyclonal antibody specifically detects Synaptotagmin 1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
Synaptotagmin 1 | |
Polyclonal | |
FITC | |
Syt1 | |
AW124717; DKFZp781D2042; FLJ42519; G630098F17Rik; P65; SV DKFZp781D2042; SV SYT; svp65; Syb1; synaptotagmin 1; synaptotagmin 1 L homeolog; synaptotagmin 2; synaptotagmin I; synaptotagmin I protein; synaptotagmin I variant b; Synaptotagmin II; Synaptotagmin XI; synaptotagmin-1; Synaptotagmin-2; SYT; Syt1; syt1.L; SYT2; SytI; SytII; XELAEV_18022019mg | |
Synthetic peptide taken within amino acid region 1-60 on SYT1 protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P21579, P21707, P46096 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20979, 25716, 6857 | |
-20° C, store in dark | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title