Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptotagmin-1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$177.00 - $450.00
Specifications
Antigen | Synaptotagmin-1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN218025UL
|
MilliporeSigma
ABN218025UL |
25 μL |
Each for $177.00
|
|
|||||
ABN2180100
|
MilliporeSigma
ABN2180100UL |
100 μL |
Each for $450.00
|
|
|||||
Description
Synaptotagmin-1 Polyclonal antibody specifically detects Synaptotagmin-1 in Rat samples. It is validated for Activity Assay, Immunocytochemistry, Immunofluorescence, Western Blotting.Specifications
Synaptotagmin-1 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Rat | |
BSA-conjugated linear peptide corresponding to the first 23 amino acids from the N-terminal vesicular domain of rat Synaptotagmin-1. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
0.5 mg/mL | |
Polyclonal | |
Purified | |
RUO | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title