Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptotagmin 12 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Synaptotagmin 12 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP169103
|
Novus Biologicals
NBP169103 |
100 μL |
Each of 1 for $436.00
|
|
Description
Synaptotagmin 12 Polyclonal specifically detects Synaptotagmin 12 in Human samples. It is validated for Western Blot.Specifications
Synaptotagmin 12 | |
Polyclonal | |
Rabbit | |
Membrane Vesicle Markers | |
SRG1, synaptotagmin XIIsynaptotagmin-XII, synaptotagmin-12, SYT11, SytXII | |
SYT12 | |
IgG | |
Affinity Purified | |
46 kDa |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
91683 | |
Synthetic peptides corresponding to SYT12 (synaptotagmin XII) The peptide sequence was selected from the N terminal of SYT12. Peptide sequence AEAREKRVPAWNAQRASTRGPPSRKGSLSIEDTFESISELGPLELMGREL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title