Learn More
Synaptotagmin 3 Mouse anti-Human, Mouse, Rat, Clone: S278-19, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527604
Catalog No. PIMA527604
Add to cart
Description
Description
Synaptotagmins are a large gene family of synaptic vesicle type III integral membrane proteins that function as regulators of both exocytosis and endocytosis and are involved in neurotransmitter secretion from small secretory vesicles. Calcium binds to Synaptotagmin I which triggers neurotransmitter release at the synapse. Synaptotagmin II is phosphorylated by WNK1 in a process that regulates calcium-dependent interactions. Synaptotagmin III is involved in calcium-dependent exocytosis of secretory vesicles in endocrine cells and neurons. Synaptotagmin IV is expressed in neuronal tissues, and has the highest mRNA levels in the hippocampus. The proximity of the Synaptotagmin IV gene to markers of several psychiatric disorders suggest an involvement of synaptotagmin IV in human disease. Synaptotagmin V is a dense-core vesicle-specific protein that regulates a specific type of calcium-regulated secretion. Synaptotagmin VI interacts with adaptor protein-2 in a calcium-independent manner. Synaptotagmin VII is widely expressed in non-neuronal tissues.
Specifications
Specifications
Synaptotagmin 3
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
O35681, P40748, Q9BQG1
SYT-3, SYT3, Synaptotagmin 3, Synaptotagmin III, SytIII, Syt III, Synaptotagmin3, SynaptotagminIII
Mouse
IgG1
100 μg
-20°C
Primary
20981, 25731, 84258
Immunocytochemistry, Immunofluorescence, Western Blot
S278-19
Unconjugated
SYT3
Liquid
Syt3
Fusion protein amino acids 442-533 (Cytoplasmic C2B domain) of mouse Synaptotagmin-3
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat