Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptotagmin 3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Synaptotagmin 3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159689
|
Novus Biologicals
NBP159689 |
100 μL |
Each for $436.00
|
|
NBP15968920
|
Novus Biologicals
NBP15968920UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Synaptotagmin 3 Polyclonal specifically detects Synaptotagmin 3 in Human samples. It is validated for Western Blot.Specifications
Synaptotagmin 3 | |
Polyclonal | |
Rabbit | |
Membrane Vesicle Markers | |
Q9BQG1 | |
84258 | |
Synthetic peptides corresponding to SYT3(synaptotagmin III) The peptide sequence was selected from the N terminal of Synaptotagmin 3. Peptide sequence VSWKLCWVPWRDKGGSAVGGGPLRKDLGPGVGLAGLVGGGGHHLAAGLGG. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
synaptotagmin III, synaptotagmin-3, SytIII | |
SYT3 | |
IgG | |
Affinity Purified | |
63 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title