Learn More
Synaptotagmin-4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 204721AP
Catalog No. 50-173-2910
Add to cart
Description
Description
May be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis. Plays a role in dendrite formation by melanocytes.
Specifications
Specifications
Synaptotagmin-4
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P40749, P50232, Q9H2B2
HsT1192, KIAA1342, Synaptotagmin 4, synaptotagmin IV, Synaptotagmin-4, SYT4, SytIV
Rabbit
IgG
150 μL
-20°C
Primary
20983, 64440, 6860
Western Blot
Unconjugated
SYT4
Liquid
Syt4
Synaptotagmin-4 Fusion Protein Ag14318
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat