Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synaptotagmin 4 Mouse anti-Human, Clone: OTI2D3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526215

Catalog No. PIMA526215


Add to cart

Description

Description

May be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis. Plays a role in dendrite formation by melanocytes.
Specifications

Specifications

Synaptotagmin 4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H2B2
Synaptotagmin-4; Synaptotagmin 4; Synaptotagmin4; Synaptotagmin IV; SytIV; SYT4; KIAA1342
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6860
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2D3
Unconjugated
SYT4
Liquid
SYT4
Full length human recombit protein of SYT4 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit