Learn More
Synaptotagmin 6 Mouse anti-Human, Mouse, Rat, Clone: S270-47, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527599
Catalog No. PIMA527599
Add to cart
Description
Description
Synaptotagmins, such as SYT6, share a common domain structure that includes a transmembrane domain and a cytoplasmic region composed of 2 C2 domains. Some synaptotagmins are involved in synaptic membrane fusion, while others have a more general function in endocytosis. For further information on synaptotagmins, see MIM 185605.
Specifications
Specifications
Synaptotagmin 6
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Q5T7P8, Q62746, Q9R0N8
SYT-6, SYT6, Synaptotagmin 6, Synaptotagmin VI, SytVI, Syt VI, Synaptotagmin6, SynaptotagminVI
Mouse
IgG1
100 μg
-20°C
Primary
148281, 54524, 60565
Immunocytochemistry, Immunofluorescence, Western Blot
S270-47
Unconjugated
SYT6
Liquid
SYT6
Fusion protein amino acids 246-333 (Cytoplasmic C2A domain) of mouse Synaptotagmin-6
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat