Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synaptotagmin 6 Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297014AF405
Description
Synaptotagmin 6 Polyclonal specifically detects Synaptotagmin 6 in Human samples. It is validated for ELISA.Specifications
Synaptotagmin 6 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 405 | |
synaptotagmin-6, sytVI | |
Produced in rabbits immunized with purified, recombinant Synaptotagmin 6 (NP_995320.1; Val359-Leu510). | |
0.1 mL | |
Membrane Vesicle Markers, Phospho Specific | |
148281 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title