Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Synaptotagmin 7 Monoclonal Antibody (N275/14)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527654

Catalog No. PIMA527654


Only null left
Add to Cart

Description

Description

1 μg/mL of MA5-27654 was sufficient for detection of Synaptotagmin-7 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 45kDa. Does not cross-react with Synaptotagmin-6 (or others). Can identify other isoforms bands at approximately 65kD. This antibody was formerly sold as clone S275-14.

This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. A similar protein in rodents mediates hormone secretion and lysosome exocytosis. In humans, expression of this gene has been associated with prostate cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Synaptotagmin 7
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
O43581, Q62747, Q9R0N7
SYT7
Fusion protein amino acids 150-239 (Cytoplasmic C2A domain) of mouse Synaptotagmin-7.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
N275/14
Unconjugated
SYT7
AI851541; B230112P13Rik; IPCA7; IPCA-7; PCANAP7; Prostate cancer-associated protein 7; Protein Syt7; synaptotagmin 7; synaptotagmin VII; Synaptotagmin-7; SYT7; SYT-7; SytVII; SYT-VII
Mouse
Protein G
RUO
54525, 59267, 9066
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.