Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Synaptotagmin 9 Mouse anti-Human, Mouse, Rat, Clone: S276-15, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527659
Catalog No. PIMA527659
Description
Description
SYT9 may be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis.
Specifications
Specifications
Synaptotagmin 9
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
O00445, P47861, Q9R0N5
SYT-9, SYT9, Synaptotagmin 9, Synaptotagmin IX, SytIX, Syt IX, Synaptotagmin9, SynaptotagminIX, FLJ45896
Mouse
IgG1
100 μg
-20°C
Primary
53420, 54309, 6861
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S276-15
Unconjugated
SYT5
Liquid
Syt5
Fusion protein amino acids 253-344 (Cytoplasmic C2B domain) of mouse Synaptotagmin-9
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat