Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI SyncorePlus Polyvap Parallel Evaporator System
SDP

SyncorePlus Polyvap allows fast evaporation of up to 96 samples in parallel to residue. In a system with the V-300 and I-300 Pro, the SyncorePlus is upgraded to a fully-automated instrument. Includes S-Condenser, V-300 vacuum pump with secondary condenser, I-300 Pro interface, and F-308 chiller.

Supplier:  BUCHI 11SQP3S000121

 View more versions of this product

Catalog No. 05-405-372


Explore available promotions
Add to Cart
Request a Quote

Description

Description

  • Save time and increase efficiency by evaporating multiple samples with volumes ranging from 0.5 mL to 500 mL
  • Compatible with BUCHI’s Polyvap glass tubes of all sizes in 4, 6, and 12 rack positions
  • Third party test tubes are compatible with 24, 48, and 96 position racks
Specifications

Specifications

50/60 Hz
Multi-Sample Evaporation
115 V
SyncorePlus Polyvap
EasyFill R-12 Polyvap
520 x 500 x 325 mm
60 to 400 rpm
12 x 125 mL
30 kg
20°C to 100°C
1250 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.