Learn More
Syndecan-1/CD138 Rat anti-Human, Alexa Fluor 594, Clone: 359103R, R&D Systems
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB2780RT100UG
Catalog No. FB2780RT100
Add to cart
This item is not returnable. View return policy
Description
Description
Syndecan-1/CD138 Monoclonal antibody specifically detects Syndecan-1/CD138 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Syndecan-1/CD138
359103R
Flow Cytometry 0.25-1 μg/106 cells
NP_002988
CD138, CD138 antigen, SDC, SDC1, SYND1heparan sulfate proteoglycan fibroblast growth factor receptor, syndecan, syndecan 1, syndecan proteoglycan 1, syndecan-1, Syndecan1
Mouse myeloma cell line NS0-derived recombinant human Syndecan-1/CD138, Gln18-Glu251, Accession No. NP_002988
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Syndecan-1/VD138 in direct ELISAs.
Flow Cytometry
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Conjugated, Purified, Supernatant
Rat
IgG1
100 μg
Primary
6382.0
Human