Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Syndecan 1 (SDC1) Human ELISA Kit Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Sandwich ELISA Kit

$494.00

Specifications

Storage Requirements 2°C to 8°C
Assay Sensitivity 14 pg/mL
Assay Range 16.38-4000 pg/mL
Test Time 1 hr. 20 min.
Sample Volume Plasma, 15 μL; Serum, 15 μL; Supernatant, 100 μL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EHSDC1 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Invitrogen™
EHSDC1
96 Tests
Pack for $494.00
Add to cart
 
Description

Description

The Human Syndecan 1 (SDC1) ELISA quantitates Hu Syndecan 1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Syndecan 1. Principle of the method The Human Syndecan 1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CD138 (Syndecan 1) is a transmembrane proteoglycan made up of one core protein and five glycosaminoglycans that can bind a variety of cytokines and modulate their activity, as well as the activity of extracellular matrix components and influence many developmental processes. CD138 is expressed mainly in differentiating keratinocytes and is transiently upregulated in all layers of the epidermis upon tissue injury. CD138 is also highly expressed on plasma cells and can be detected even on fibroblasts, vascular smooth muscle cells and endothelial cells. Up-regulation and down-regulation of CD138 on the cell surface often correlates with the gain of cancerous characteristics. Serum levels of the shedded soluble sCD138 are used as a prognostic factor of carcinogenesis. CD138 is expected to play a role in cell adhesion, cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins. Further, CD138 is expressed on the surface of pre-B cells and plasma cells but is absent from mature B cells, and is lost from the apoptotic myeloma cells. Altered CD138 expression has been detected in several different tumor types. While several transcript variants may exist for the CD138 gene, the full-length natures of only two have been described to date which represent the major variants and encode the same protein.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

2°C to 8°C
16.38-4000 pg/mL
Plasma, 15 μL; Serum, 15 μL; Supernatant, 100 μL
<10%
14 pg/mL
1 hr. 20 min.
<12%
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit