Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syndecan-4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50
Specifications
Antigen | Syndecan-4 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017971
![]() |
Novus Biologicals
NB110415510.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NB11041551
![]() |
Novus Biologicals
NB11041551 |
0.1 mg | N/A | N/A | N/A | ||||
Description
Syndecan-4 Polyclonal specifically detects Syndecan-4 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Syndecan-4 | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4 | |
SDC4 | |
IgG | |
Affinity purified | |
22 kDa |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Chicken, Mouse, Rat, Human | |
P31431 | |
6385 | |
Synthetic peptide surrounding amino acid 184 of human Syndecan-4. | |
Primary | |
Store at -20 degreesC. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title