Learn More
Syndecan-4 Rat anti-Human, Alexa Fluor™ 750, Clone: 336304, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB29181S100UG
Catalog No. FB29181S100
Add to cart
This item is not returnable. View return policy
Description
Description
Syndecan-4 Monoclonal antibody specifically detects Syndecan-4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Syndecan-4
336304
Flow Cytometry 0.25-1 μg/106 cells
P31431
amphiglycan, MGC22217, ryudocan, Ryudocan core protein, SDC4, SYND4ryudocan amphiglycan, syndecan 4, syndecan 4 (amphiglycan, ryudocan), syndecan proteoglycan 4, syndecan-4, Syndecan4
Mouse myeloma cell line NS0-derived recombinant human Syndecan-4, Ser20-Glu145, Accession # P31431
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human Syndecan-4 in direct ELISAs.
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Rat
IgG2a
100 μg
Primary
6385
Human