Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synembryn-A/Ric8A Mouse anti-Rat, Human, Clone: 3E1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS1241
Description
Detect Synembryn-A/Ric-8A using this mouse monoclonal Anti-Synembryn-A/Ric8A antibody, clone 3E1 validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Specifications
Synembryn-A/Ric8A | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Ric8a | |
A full-length recombinant Ric-8A protein from rat. | |
100 μg | |
Signaling | |
Clone 3E1 detects Ric-8A in human brain. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2a κ |
Western Blot, Immunohistochemistry (Paraffin) | |
3E1 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Rat, Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction