Description
This unique system is based on an embedded Windows XP processor and provides a powerful image capture and storage device. A very large touch screen gives instant access to a range of powerful functions in this feature packed system. The camera and 5 position filter wheel are fully motorised. An internal hard disk and a USB port for saving images to a memory stick provide an excellent choice for the archiving of images in different file formats. The system can also be adapted for networking using an ethernet connection. This system comes complete with Syngene's powerful GeneTools analysis software.
Applications
- Electrophoresis gels - Ethidium Bromide, Coomassie Blue, Silver Stain, SYBR™ Gold, SYBR Green, SYBR Safe, GelStar™, SYPRO™Red, SYPRO Ruby, SYPRO Orange, Fluorescein, Rhodamine Red™, Texas Red™, Pro-Q™ Diamond, Deep Purple™
- Agar plates
- Cells in flasks
- Autoradiographs
- Membranes
- Spots and slot blots
- TLC plates
- Microtitre plates
- Macro-arrays
Features - Compact darkroom, small footprint taking up minimal laboratory bench space
- Slide in and out transilluminator and drop down white light table, multi application use
- Integral computer, no additional PC required
- Large touch screen, full intuitive touch control of all functions with large image view
- Save images in TIFF, BMP or JPEG format, simple to download to any PC or Mac
Specifications
Specifications
| For Use With (Application) | GeneGenius, MultiGenius, ChemiGenius |
| Product Type | Filter Long Pass |
| For Use With (Equipment) | GeneGenius, MultiGenius, ChemiGenius |
| Wavelength | 515-599nm |