Learn More
Description
- For use in GBOX and Gbox Mini systems 7 position filter wheel.
Specifications
Specifications
| For Use With (Application) | Chemi Systems and 7 Position Filter Wheel |
| For Use With (Equipment) | Gbox and Gbox Mini |
| Wavelength | 572 to 625 nm |
| Product Type | Filter UV |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

