Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ Emission Filter
GREENER_CHOICESDP

Catalog No. 01257198 Shop All Synoptics, Inc. Products
Click to view available options
Wavelength:
605 ±10 nm
655 nm, 633 to 675 nm range
705 ±10 nm

Catalog No. 01-257-198

Supplier: Syngene™ FILT655GX

Only null left
Add to Cart
Add to Cart

Syngene™ Emission Filter works in conjuction with the Syngene Gbox and Gbox Mini Systems.

  • For use in GBOX and Gbox Mini systems 7 position filter wheel.

Specifications

Intensity Fixed
Wavelength 655 nm, 633 to 675 nm range
For Use With (Equipment) Gbox and Gbox Mini
Product Type Filter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.