Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYNM Mouse anti-Human, Clone: OTI3D10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene CF806549
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Type-VI intermediate filament (IF) which plays an important cytoskeletal role within the muscle cell cytoskeleton. It forms heteropolymeric IFs with desmin and/or vimentin, and via its interaction with cytoskeletal proteins alpha-dystrobrevin, dystrophin, talin-1, utrophin and vinculin, is able to link these heteropolymeric IFs to adherens-type junctions, such as to the costameres, neuromuscular junctions, and myotendinous junctions within striated muscle cells.Specifications
SYNM | |
Monoclonal | |
Unconjugated | |
SYNM | |
DMN, SYN | |
Mouse | |
Affinity Chromatography | |
RUO | |
23336 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3D10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O15061 | |
SYNM | |
Human recombit protein fragment corresponding to amino acids 329-580 of human SYNM produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction