Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SYNM Monoclonal Antibody (OTI7H6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526802
Description
SYNM Monoclonal antibody specifically detects SYNM in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SYNM | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15061 | |
SYNM | |
Human recombinant protein fragment corresponding to amino acids 329-580 of SYNM produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7H6 | |
Unconjugated | |
SYNM | |
4930412K21Rik; AI852401; Desmuslin; DMN; E130104F11; KIAA0353; SYN; synemin; synemin alpha; synemin beta; synemin, intermediate filament protein; synm | |
Mouse | |
Affinity Chromatography | |
RUO | |
23336 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction