Learn More
Syntaxin 18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160131AP
Catalog No. 50-173-0642
Add to cart
Description
Description
Syntaxins, a family of proteins involved in the fusion of synaptic vesicles with the plasma membrane, display broad tissue distribution and contain carboxy-terminal hydrophobic domains that direct themselves to their respective intracellular compartments. Syntaxin 18 mainly localizes to the endoplasmic reticulum (ER), and functions in transport between the ER and Golgi. Syntaxin 18 also plays a role in ER-mediated phagocytosis, possibly by regulating the specific fusion of the ER and plasma or phagosomal membranes.
Specifications
Specifications
Syntaxin 18
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q68FW4, Q8VDS8, Q9P2W9
STX18, syntaxin 18, Ufe1
Rabbit
IgG
150 μL
-20°C
Primary
360953, 53407, 71116
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
STX18
Liquid
Stx18
Syntaxin 18 Fusion Protein Ag8777
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat