Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syntaxin 3 Rabbit anti-Human, Alexa Fluor 594, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP300343AF594

Catalog No. NB065668


Add to cart

Description

Description

Syntaxin 3 Polyclonal antibody specifically detects Syntaxin 3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Syntaxin 3
Alexa Fluor 594
50mM Sodium Borate
FLJ30906, STX3Asyntaxin 3A, syntaxin 3, syntaxin-3
Produced in rabbits immunized with purified, recombinant Syntaxin 3 (NP_004168.1; Met1-Asn289).
Affinity purified
RUO
Primary
6809
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit