Learn More
Syntaxin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155561AP
Catalog No. 50-173-0314
Add to cart
Description
Description
The gene is a member of the syntaxin family. The encoded protein is targeted to the apical membrane of epithelial cells where it forms clusters and is important in establishing and maintaining polarity necessary for protein trafficking involving vesicle fusion and exocytosis. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
Syntaxin 3
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q08849, Q13277, Q64704
STX3, STX3A, syntaxin 3
Rabbit
IgG
150 μL
-20°C
Primary
20908, 6809, 81802
Immunoprecipitation, Western Blot
Unconjugated
STX3
Liquid
Stx3
Syntaxin 3 Fusion Protein Ag7925
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat