Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Syntaxin 5 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | Syntaxin 5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB023880
|
Novus Biologicals
NBP294678100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB023881
|
Novus Biologicals
NBP29467820UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Syntaxin 5 Polyclonal antibody specifically detects Syntaxin 5 in Rat samples. It is validated for Western BlotSpecifications
Syntaxin 5 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
6811 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat | |
STX5ASED5, syntaxin 5, syntaxin 5A, syntaxin-5 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 55-355 of human STX5 (NP_003155.2). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title