Learn More
Synthware™ Chromatography Column with Spherical Joint, Reservoir, Fritted Disc and PTFE Stopcock
For flash chromatography
Supplier: Synthware™ C383240C
Description
- General purpose chromatography column with reservoir, PTFE stopcock and top 35/20 spherical joint
- With coarse porosity fritted disc to support column packing
- Heavy-wall design increases safety
Specifications
26 mm | |
35/20 | |
457 mm |
500 mL | |
2 mm bore |