Learn More
Synthware™ Chromatography Column with Spherical Joint, Reservoir and PTFE stopcock
GREENER_CHOICESDP
For flash chromatography
Supplier:  Synthware™ C382625
 View more versions of this product
Catalog No. 31-500-942
Add to Cart
Description
Description
  • General purpose chromatography column with reservoir, PTFE stopcock and top 35/20 spherical joint
  • Heavy-wall design increases safety
Specifications
Specifications
20 mm
2 mm bore
254 mm
35/20
250 mL
Product Suggestions
Product Suggestions