Learn More
Synthware™ Chromatography Column with Standard Taper Joint, Reservoir and PTFE Stopcock
For flash chromatography
Supplier: Synthware™ C374620
Description
- General purpose chromatography column with reservoir, PTFE stopcock and top standard taper outer joint
- Heavy-wall design increases safety
Specifications
40 mm | |
24/40 | |
2 mm bore | |
203 mm |