Learn More
Synthware™ Chromatography Reservoir, with Spherical Joints
GREENER_CHOICESDP
With spherical joints
Supplier:  Synthware™ C193502
 View more versions of this product
Catalog No. 31-500-839
Add to Cart
Description
Description
  • Used for flash chromatography
Specifications
Specifications
250 mL
Top Socket Joint: 35/20, Bottom Ball Joint: 35/20
Product Suggestions
Product Suggestions