Learn More
Synthware™ Chromatography Columns, Modified
Short column with lower 24/40 joint and hose connection for vacuum
$144.50 - $301.50
Specifications
Type Chromatography Column, Modified
Material Borosilicate Glass 3.3
View More Specs
Products 6
Catalog Number Mfr. No. Diameter (Metric) Outer Length (Metric) Description Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Outer Length (Metric) Description Price Quantity & Availability  
31-500-964
View Documents
Synthware™
F234020C
40 mm 200 mm Coarse
Each for $144.50
Add to cart
31-500-965
View Documents
Synthware™
F234030C
40 mm 300 mm Coarse
Each for $161.90
Add to cart
31-500-967
View Documents
Synthware™
F236040C
60 mm 400 mm Coarse
Each for $242.50
Add to cart
31-500-966
View Documents
Synthware™
F234030M
40 mm 300 mm Medium
Each for $161.90
Add to cart
31-500-968
View Documents
Synthware™
F238530C
90 mm 300 mm Coarse
Each for $274.50
Add to cart
31-500-969
View Documents
Synthware™
F238540C
90 mm 400 mm Coarse
Each for $301.50
Add to cart
Description
Description
Synthware Chromatography funnels are short, with a lower 24/40 inner joint and feature a sealed in drip tip and hose connection.
Specifications
Specifications
Chromatography Column, Modified
Borosilicate Glass 3.3