Kemtech America™ Synthware™ Concentrator Tube, Graduated

For use with Kuderna-Danish evaporative set-ups

Manufacturer: Kemtech America™ C291910

 View more versions of this product

Catalog No. 31-501-088

  Check Availability
Add to Cart

    Specifications

    Type Graduated Concentrator Tube
    Item Description Subdivisions: 0 to 1mL in 0.1; 2 to 10mL in 1.0
    Joint Type Joint Size: 19/22
    Height (Metric) 100mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit