Learn More
Synthware™ Crystallizing Dish with Spouts
With spouts
$24.67
Specifications
Description Crystallizing Dish
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Height (Metric) Price Quantity & Availability  
31-501-012
View Documents
Synthware™
D290170
900 mL 45 mm
Each for $24.67
Add to cart
Description
Description
  • Top of each dish is reinforced and fire polished to reduce chipping
Specifications
Specifications
Crystallizing Dish