Learn More
Synthware™ Distillation Receiver, Cow Type, with Hose Connection
With hose connection for application of vacuum
Supplier: Synthware™ D681922
Description
- With three receivers spaced 40 deg. apart at 105 deg. angle between top outer joint and lower inner drip tip joints
- All joints of same size
Specifications
Cow Type Distillation Receiver | |
19/22 |