Learn More
Synthware™ Erlenmeyer Flasks, Threaded
For microscale applications
Supplier: Synthware™ F501025
Description
- With 14/20 threaded joint
- Supplied with one holed Nylon compression cap, one Viton O-ring and one PTFE-faced silicone rubber septum
Specifications
Erlenmeyer Flask | |
25 mL | |
14/10 |