Kemtech America™ Synthware™ Evaporation Flasks
For controlled evaporation of foaming solvents such as toluene or xylene on rotary evaporator
Manufacturer: Kemtech America™ F794000
 View more versions of this product
Catalog No. 31-501-186
  Check Availability
Add to Cart
    Specifications
    Capacity (Metric) 1000mL
    Product Type Evaporation Flask
    Standard Taper Joint Number Center 24/40
  • Description & Specifications
Related Products
Product Content Feedback