Kemtech America™ Synthware™ Filter Funnels with Fritted Disc and Glass Stopcock
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
Manufacturer: Kemtech America™ F973014M
 View more versions of this product
Catalog No. 31-501-608
  Check Availability
Add to Cart
    Specifications
    Capacity (Metric) 60mL
    Diameter (Metric) Disc Frit: 30mm
    Joint Number 14/20
    Porosity Medium
    Product Type Filter Funnels with Fritted Disc and Glass Stopcock
  • Description & Specifications
Related Products
Product Content Feedback