Learn More
Synthware™ Filter Funnels with Perforated Glass Plate
Used in conjunction with vacuum source to filter and collect fluids
Manufacturer:  Synthware™ F932475
 View more versions of this product
Catalog No. 31-501-461
Add to cart
Description
Description
  • 24/40 standard taper inner joint
  • Hose connector for connection to vacuum source
  • Glass plate O.D.: 24mm
Specifications
Specifications
24 mm Plate
Filter Funnel with Perforated Glass Plate
75 mm