Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Filter Tubes with Fritted Disc
Used to filter particulates from substance
Supplier: Synthware™ F351430M
Description
- Standard taper inner joint
- Sealed in glass fritted disc of listed porosity
- Hose connector allows connection to a vacuum source
Specifications
Approximate frit O.D.: 30 mm, Porosity: Medium | |
Medium | |
Approximate frit O.D.: 30 mm |
Two 2 mm glass stopcocks | |
14/20 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction