Learn More
Synthware™ Filter Tubes with Fritted Disc
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
$98.77 - $113.20
Specifications
Description Filter tube with fritted disc
View More Specs
Products 6
Catalog Number Mfr. No. Standard Taper Joint Item Description Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint Item Description Price Quantity & Availability  
31-501-556
View Documents
Synthware™
F701420C
14/20 Approximate Frit O.D.: 20mm, Coarse porosity Each for $98.77
Add to cart
31-501-557
View Documents
Synthware™
F701420M
14/20 Approximate Frit O.D.: 20mm, Medium porosity Each for $102.89
Add to cart
31-501-560
View Documents
Synthware™
F702440C
24/40 Approximate Frit O.D.: 36mm, Coarse porosity Each for $113.20
Add to cart
31-501-561
View Documents
Synthware™
F702440M
24/40 Approximate Frit O.D.: 36mm, Medium porosity Each for $113.20
Add to cart
31-501-558
View Documents
Synthware™
F701430C
14/20 Approximate Frit O.D.: 30mm, Coarse porosity Each for $104.94
Add to cart
31-501-559
View Documents
Synthware™
F701430M
14/20 Approximate Frit O.D.: 30mm, Medium porosity Each for $109.05
Add to cart
Description
Description
  • With standard taper inner joints
  • Available with either coarse or medium porosity fritted disc
Specifications
Specifications
Filter tube with fritted disc