Learn More
Synthware™ Two Neck Pear Shaped Flasks
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
Manufacturer:  Synthware™ F341125
 View more versions of this product
Catalog No. 31-501-203
Add to cart
Description
Description
  • Standard taper inner joint at each end
  • Available with either coarse or medium porosity fritted disc
  • With two 2 mm glass stopcocks having constant pressure retention devices
Specifications
Specifications
Two neck pear shaped flask
25 mL
Center: 14/20; Side: 14/20