Learn More
Synthware™ Two Neck Pear Shaped Flasks
Used for filtration followed by recrystallization on opposite side of disc of air sensitive compounds
$42.40 - $55.40
Specifications
Description Two neck pear shaped flask
View More Specs
Products 7
Catalog Number Mfr. No. Standard Taper Joint Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint Capacity (Metric) Price Quantity & Availability  
31-501-200
View Documents
Synthware™
F341105
Center: 14/20; Side: 14/20 5 mL
Each for $42.40
Add to cart
31-501-201
View Documents
Synthware™
F341110
Center: 14/20; Side: 14/20 10 mL
Each for $42.90
Add to cart
31-501-202
View Documents
Synthware™
F341115
Center: 14/20; Side: 14/20 15 mL
Each for $43.50
Add to cart
31-501-203
View Documents
Synthware™
F341125
Center: 14/20; Side: 14/20 25 mL
Each for $43.75
Add to cart
31-501-205
View Documents
Synthware™
F341100
Center: 14/20; Side: 14/20 100 mL
Each for $45.15
Add to cart
31-501-204
View Documents
Synthware™
F341150
Center: 14/20; Side: 14/20 50 mL
Each for $43.20
Add to cart
31-501-206
View Documents
Synthware™
F341200
Center: 14/20; Side: 14/20 200 mL
Each for $55.40
Add to cart
Description
Description
  • Standard taper inner joint at each end
  • Available with either coarse or medium porosity fritted disc
  • With two 2 mm glass stopcocks having constant pressure retention devices
Specifications
Specifications
Two neck pear shaped flask